Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203697

Sigma-Aldrich

BPDQ

A cell-permeable, reversible, ATP-competitive, potent and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR; IC₅₀ = 120 pM).

Synonym(s):

BPDQ, 4-[(3-Bromophenyl)amino]-6,7-diaminoquinazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12BrN5
CAS Number:
Molecular Weight:
330.18
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, reversible, ATP-competitive, potent and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR; IC50 = 0.12 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGFR
Product competes with ATP.
Reversible: yes
Target IC50: 120 pM against EGFR

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). This product is stable for up to 3 months at -20°C.

Other Notes

Rewcastle, G.W., et al. 1996. J. Med. Chem. 39, 918.
Rewcastle, G.W., et al. 1995. J. Med. Chem. 38, 3482.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service