203351-M
Blasticidin S, Hydrochloride, Streptomyces sp., Sterile-Filtered Aqueous Solution, Cell Culture-Tested
sterile; sterile-filtered, suitable for cell culture
Synonym(s):
Blasticidin S, Hydrochloride, Streptomyces sp., Sterile-Filtered Aqueous Solution, Cell Culture-Tested
About This Item
Recommended Products
sterility
sterile; sterile-filtered
Quality Level
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
storage temp.
−20°C
General description
Warning
Physical form
Reconstitution
Other Notes
Kimura, M., et al. 1994. Mol. Gen. Genet.242, 121.
Kojima, N., et al. 1994. J. Biol. Chem.269, 30451.
Kudo, T., et al. 1994. J. Immunol. Methods177, 17.
Sutoh, K. 1993. Plasmid30, 150.
Izumi, M., et al. 1991. Exp. Cell Res.197, 229.
Yamaguchi, H., et al. 1965. J. Biol. Chem.57, 667.
Takeuchi, S., et al. 1958. J. Antibiot.11, 1.
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service