Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.88025

Supelco

Test Kit for water determination

suitable for Karl-Fischer titration, for use with AQUASTAR®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601

technique(s)

Karl-Fischer titration: suitable

Quality Level

compatibility

for use with AQUASTAR®

Storage temp.

15-25°C

General description

according to Karl Fischer Aquastar®

Analysis Note

Extent of testing: passes test

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service