Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

17-10121

Sigma-Aldrich

ChIPAb+ Acetyl-Histone H4 (Lys12) - ChIP Validated Antibody and Primer Set, rabbit monoclonal

from rabbit

Synonym(s):

H4K12Ac, Histone H4 (acetyl K12)

Sign Into View Organizational & Contract Pricing

Select a Size

25 ASSAYS
$580.00

$580.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ASSAYS
$580.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.52

$580.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

clone

monoclonal

species reactivity

vertebrates, human

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
cell based assay: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Related Categories

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Acetyl-Histone H4 (Lys12) set includes the Acetyl-Histone H4 (Lys12) antibody, a negative control normal rabbit IgG, and qPCR primers which amplify a 213 bp region of human GAPDH coding region. The Acetyl-Histone H4 (Lys12) and negative controls are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of Acetyl-Histone H4 (Lys12) associated chromatin.

Specificity

This antibody detects histone H4 acetylated at Lys12.
Wide range of cross-reactivity expected based on sequence homology.

Immunogen

Epitope: Unknown
Peptide corresponding to Histone H4 containing the sequence [GLG-AcK-GGA] on which Lys12 is acetylated.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using either 2 µL of Negative Control Supernatant, or 2 µL of Anti-Acetyl-Histone H4 (Lys12) and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of Acetyl-Histone H4 (Lys12)-associated DNA fragments was verified by qPCR using ChIP Primers, human GAPDH Coding Region as a positive locus, and a gene desert region as a negative locus. (Figure 2). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Western Blot Analysis:
Representative lot data.
Lysates from HeLa cells untreated or sodium butyrate treated (Lanes 1 and 2 respectively) were resolved probed with anti-acetyl-Histone H4 (Lys12) (1:1,000). Arrow indicates Acetyl-Histone H4 (Lys12).
Arrow indicates Acetyl-Histone H4 (Lys12) (~11 kDa) (Figure 3).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Chromatin Biology
This ChIPAb+ Acetyl-Histone H4 (Lys12) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per set. Recommended use: 2 μL of antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa S3 cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using either 2 µL of Negative Control Supernatant, or 2 µL of Anti-Acetyl-Histone H4 (Lys12) and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of Acetyl-Histone H4 (Lys12)-associated DNA fragments was verified by qPCR using ChIP Primers, human GAPDH promoter (Figure 1).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

11kDa

Physical form

Anti-Acetyl-Histone H4 (Lys12) (rabbit monoclonal), . One vial containing 50 µL of rabbit monoclonal IgG cell culture supernatant in 0.1% sodium azide. Store at -20°C.
Negative Control Supernatant, . One vial containing 100 µL of cultured supernatant in 0.05% sodium azide. Store at -20°C.
ChIP Primers, human GAPDH coding region,. One vial containing 75 μL of 5 μM each primer specific for human GAPDH coding region.
Store at -20°C.
FOR: GGC TCC CAC CTT TCT CAT CC
REV: GGC CAT CCA CAG TCT TCT GG
Format: Purified
Protein A purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Includes negative control supernatant and primers specific for human GAPDH coding region.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Ashbrook et al.
Clinical and experimental immunology, 180(3), 520-530 (2015-01-27)
Citrate, a central component of cellular metabolism, is a widely used anti-coagulant due to its ability to chelate calcium. Adenosine triphosphate (ATP)-citrate lyase, which metabolizes citrate, has been shown to be essential for inflammation, but the ability of exogenous citrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service