Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46686

Millipore

Choc.Agar LTH 90mm ICR+

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

material

polystyrene plate

Quality Level

agency

non-compendial

sterility

sterile; γ-irradiated by SER-TAIN process

form

solid

feature

2D bar code
ready-to-use

D

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring
microbiology
pharmaceutical

suitability

anaerobic bacteria
bacteria
molds
yeasts

General description

Application: Environmental monitoring for isolators & clean rooms, Active or passive air monitoring, Fingerprint, Detection of fastidious microorganisms

Packaging: 90 mm settle plate with safety lock (20 or 120 plates per box)

Synonym: Chocolate Agar with neutralizer, Chocolate Agar with Lecithin, Tween and Histidin

Legal Information

SER-TAIN is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service