Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46346

Millipore

Mossel Broth (EE Broth)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703

form

liquid

feature

ready-to-use

pH

7.2 ( in H2O)

application(s)

food and beverages
industrial qc
microbiology

Storage temp.

20-25°C

suitability

enterobacteriaceae

Application

Enrichment of Enterobacteriaceae (Enterobacter)

Packaging

10 ml in 17 ml tube with red cap (20 tubes per box)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service