Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.44305

Millipore

Thermal Printer

for use with APC Ergo Touch Pro 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

packaging

pkg of 1 kit

application(s)

air monitoring
pharmaceutical

compatibility

for use with APC Ergo Touch Pro 2

storage temp.

no temp limit

General description

942753 (APC ErgoTouch Pro 2)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service