Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.44147

Millipore

Thermal Printer Paper

for use with APC SmartTouch

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
44111506

compatibility

for use with APC SmartTouch

storage temp.

room temp

General description

192074 (APC SmartTouch)

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service