Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

144133

Millipore

Power Supply

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

agency

non-compendial

application(s)

pharmaceutical

compatibility

for use with APC Ergo Touch Pro 2

shipped in

ambient


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service