Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.44099

Millipore

Agar Strips C

MacConkey Agar, Coliform bacteria, HYCON®

Synonym(s):

Hycon Ready-to-use Agar Strips, MAC Agar Strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113035

Quality Level

sterility

non-sterile

form

plate (ready-to-use)

feature

ready-to-use

packaging

pkg of 25 units

manufacturer/tradename

HYCON®

pH

7.1 ( in H2O)

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical

compatibility

for use with RCS® Air Samplers

storage temp.

2-8°C

suitability

coliforms

General description

Agar Strips C are ready-to-use culture media for assessment of airborne coliforms and E. coli with RCS® Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments. MacConkey agar is filled in a flexible backing film. MacConkey Agar is a weak selective medium for isolation of coliforms and E. coli. Bile salts and crystal violet largely inhibit the growth of the gram-positive microbial flora. Lactose and the pH indicator neutral red are used to detect lactose degradation.

Application

Agar Strips are used for monitoring of ambient air and compressed gases with the RCS® Air Samplers

Analysis Note

Bacteria degrading lactose to acids grow in pink to red colored colonies. Additionally, Escherichia coli and other acid forming bacteria will show a zone of precipitated bile salt around the colonies. Bacteria not degrading Lactose will grow colorless.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service