Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

144077

Millipore

APC SmartTouch Download Utility Software

for use with APC SmartTouch

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
Product 144077 is not currently sold in your country. Contact Technical Service

compatibility

for use with APC SmartTouch

shipped in

ambient

storage temp.

room temp

General description

942848 (APC SmartTouch)
The software enables data transfer between APC SmartTouch Airborne Particle Counters and computers, includes tools to create configuration data and organize sampling data. It is included in the scope of delivery of the instrument.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service