Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

14-1055

Sigma-Aldrich

Human recombinant histone H2AX-H2B dimers

Human recombinant histone H2AX-H2B dimers expressed in E Coli. as fusion proteins with either an N-terminal FLAG or 6xHis epitopes & purified by Fast Protein Liquid Chromatography (FPLC).

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.32
Product 14-1055 is not currently sold in your country. Contact Technical Service

biological source

Escherichia coli

recombinant

expressed in E. coli

assay

>98% (SDS-PAGE)

manufacturer/tradename

Upstate®

concentration

1 μg/μL

technique(s)

activity assay: suitable

shipped in

dry ice

General description

Human recombinant histone H2AX-H2B dimers expressed in E Coli. as fusion proteins with either an N-terminal FLAG® or 6xHis epitopes, and purified by Fast Protein Liquid Chromatography (FPLC)
Product Source: Human
The core histones, H2A, H2B, H3, and H4, form a central octameric structure that binds approximately 147 bp of DNA to form a nucleosome, the basic unit of chromatin. The H2AX histone variant replaces conventional H2A in a subset of nucleosomes. H2AX plays a central role in DNA repair and genome stability. It prevents progression of the cell cycle in response DNA damage by ionizing radiation, and facilitates repair of double-stranded breaks in DNA. Phosphorylation of H2AX at serine 140 is required for its function as a hub for numerous signaling proteins involved in DNA damage and repair. The effects of histone variants and histone modifications can be deciphered by applications such as chromatin immunoprecipiation (ChIP), which requires high quality chromatin and essential tools, such as histone proteins and appropriate enzymes, to assemble chromatin in vitro.

Application

Research Category
Epigenetics & Nuclear Function
Suitable for in vitro chromatin assembly and enzyme activity assays

Physical form

In buffer containing 20 mM Tris-Cl pH 7.9, 1 mM EDTA, 1 M NaCl, 0.5 mM PMSF and 1 mM DTT

Storage and Stability

Stable at -80ºC for 1 year from date of receipt.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siyang Wu et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 9(8), e2104055-e2104055 (2022-01-21)
DNA repair confers the resistance of tumor cells to DNA-damaging anticancer therapies, while how reprogrammed metabolism in tumor cells contributes to such process remains poorly understood. Pyruvate kinase M2 isoform (PKM2) catalyzes the conversion of phosphoenolpyruvate to pyruvate and regulates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service