Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

124028-M

Sigma-Aldrich

Akt Inhibitor XI - Calbiochem

Synonym(s):

3-Formylchromone thiosemicarbazone, Cu(II)Cl₂ Complex, FPA-124

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

assay

≥98% (elemental analysis)

form

solid

color

green

solubility

DMSO: 2 mg/mL
H2O: 5 mg/mL

General description

A cell-permeable copper complex (Cu2+/Cu+ redox couple in the range of +0.28 to +0.35 V) that interacts with both the PH and the kinase domains of Akt and potently inhibits its kinase activity (IC50 = 100 nM). Inhibits tumor growth both in cultured cells in vitro (IC50 ranges from 10 to 34 μM) and in mice in vivo (25 mg/kg, iv) without any apparent adverse effect to the animals.
The Akt Inhibitor XI controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Biochem/physiol Actions

Target IC50:100 nM against Akt

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 1 month at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivek Barve et al.
Journal of medicinal chemistry, 49(13), 3800-3808 (2006-06-23)
The synthesis and characterization of Schiff base derivatives of 3-formylchromone 3-6 (FPA-120 to FPA-123), the minimal biologically active structural motif of soy isoflavone, genistein, and their copper(II) complexes 7-10 (FPA-124 to FPA-127) are reported here. These copper complexes possess distorted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service