1190-OP
OmniPur® Acrylamide: Bis-acrylamide 19:1 Premixed Powder
About This Item
form
crystalline solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
shipped in
ambient
storage temp.
10-30°C
General description
Specifications
DNase: None detected
Protease: None detected
RNase: None detected
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral
target_organs
Peripheral nervous system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service