Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.18313

Supelco

Cetyl trimethylammonium hydrogen sulfate

for ion pair chromatography LiChropur

Synonym(s):

Cetyl trimethylammonium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H43NO4S
CAS Number:
Molecular Weight:
381.61
MDL number:
UNSPSC Code:
12352107
EC Index Number:
269-286-4

Quality Level

assay

≥99% (acidimetric)

manufacturer/tradename

LiChropur

technique(s)

ion pair chromatography: suitable

loss

≤2% loss on drying, 80°C, vacuum

transmittance

220 nm, ≥70%
220 nm, ≥90%
250 nm, ≥98%
(0.005 M; 1 cm in H2O)

pH

≤2 (20 °C, 100 g/L in H2O)

suitability

passes test for identity (IR)

InChI

1S/C19H42N.H2O4S/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;1-5(2,3)4/h5-19H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

UCRJJNVFJGKYQT-UHFFFAOYSA-M

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service