Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.17097

Millipore

Profibus Module

Main board for use with MAS-100 Iso NT® with ProfiBus Interface

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$3,200.00

$3,200.00


Check Cart for Availability


Select a Size

Change View
1 UNIT
$3,200.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

$3,200.00


Check Cart for Availability

material

aluminum hardware

agency

non-compendial

description

interface

form

pieces

application(s)

air monitoring

compatibility

for use with MAS-100 Iso NT®

shipped in

ambient

storage temp.

no temp limit

General description

Profibus Module is an additional accessory for MAS-100 ISO NT® air samplers. MAS-100 ISO NT® is designed to monitor air microbiology in isolators and RABS. Like all other MAS-100® air samplers, it′s based on the Andersen impaction principle. All electronic and moving parts are located outside the critical zone, whilst one stainless steel sampling head for standard 90 to 100 mm Petri dishes collects samples in the containment zone. The Profibus module provides an interface that is used to communicate with the PC Software and Profibus Master software and tools.

Application

MAS-100 ISO NT® Profibus Module is an additional accessory for Air Sampler that provides interface that is used to communicate with the PC and Profibus Master Software. MAS-100 ISO NT® is used extensively in Pharmaceutical industries.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service