Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14667

Supelco

PC cable

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43212107

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 30
for use with Spectroquant® Nova 400
for use with Spectroquant® Nova 60
for use with Spectroquant® SQ 200
for use with Spectroquant® SQ 300

storage temp.

2-30°C

General description

(for serial interface) NOVA 30, NOVA 60, NOVA 400, SQ 200, SQ 300 Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service