Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14421

Supelco

Color Test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

storage temp.

15-25°C

Quality Level

General description

The MQuant® Color test (formally MColortest®) is used for the visual comparison of yellow tinted waters with simulated platinum-cobalt standards according to Hazen.

Method
The color of the liquid under investigation is measured semiquantitatively by visual comparison of the measurement solution with the color fields of a color card. The result obtained when measuring an unfiltered sample corresponds to the “apparent color”, which is caused by dissolved substances and suspended matter. When the sample is filtered prior to the measurement, the result corresponds to the “true color”, which is caused only by dissolved substances. The two results are identical in the case of clear samples. The color of water samples depends strongly on the pH. When measuring the color, it is hence advisable to also determine the pH of the water sample under investigation (e.g. with MQuant® pH indicator strips). The unique brilliance of the print and their fine color graduation allow precise analyses.

This test is suitable for testing the color in
  • Surface water
  • Drinking water
  • Water in the water-treatment process
  • Wastewater
  • Electroplating wastewater
  • Inorganic liquids (e. g. salt solutions)
  • Organic liquids (e. g. solvents)
  • Beverages
The test is used for the visual comparison of yellow tinted waters with simulated platinum-cobalt standards according to Hazen.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service