Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.11435

Millipore

DHL agar acc. to SAKAZAKI

for the detection and isolation of pathogenic Enterobacteriaceae

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

form

medium granules (dehydrated (DCM))

technique(s)

microbiological culture: suitable

pH

7.0-7.4 (25 °C, 64 g/L in H2O)

solubility

64 g/L

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

enterobacteriaceae

General description

for the detection and isolation of pathogenic Enterobacteriaceae

Analysis Note

Appearance (clearness)clearAppearance (colour)redSolidification behaviour (2 hrs., 45 °C)liquidpH-value (25 °C)7.0 - 7.4Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031))good to very goodGrowth (Salmonella enteritidis ATCC 13076 (WDCM 00030))good to very goodGrowth (Enterococcus faecalis ATCC 11700)none to poorGrowth (Staphylococcus aureus ATCC 25923 (WDCM 00034))noneGrowth (Bacillus cereus ATCC 11778 (WDCM 00001))noneGrowth (Shigella flexneri ATCC 12022 (WDCM 00126))fair to very goodColony colour (Escherichia coli ATCC 25922 (WDCM 00013))redColony colour (Klebsiella pneumoniae ATCC 10031)pinkColony colour (Salmonella typhimurium ATCC 14028 (WDCM 00031))colourlessColony colour (Salmonella enteritidis ATCC 13076 (WDCM 00030))colourlessColony colour (Shigella flexneri ATCC 12022 (WDCM 00126))colourlessColonies (Proteus mirabilis ATCC 14153)colourless; brownish zoneBlack centre (Salmonella typhimurium ATCC 14028 (WDCM 00031))+Black centre (Salmonella enteritidis ATCC 13076 (WDCM 00030))+Precipitate (Escherichia coli ATCC 25922 (WDCM 00013))+Incubation: 24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service