Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

109789-M

Ethernet Module

Main board for use with MAS-100 Iso NT®/ MAS-100 Iso MH® instruments with Ethernet interface

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
Pricing and availability is not currently available.

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100 ISO MH® Microbial Air Sampler
for use with MAS-100 ISO NT® Microbial Air Sampler

storage temp.

no temp limit

General description

Ethernet Module is an additional accessory for MAS-100 ISO NT® air samplers. MAS-100 ISO NT® is designed to monitor air microbiology in isolators and RABS. Like all other MAS-100® air samplers, it′s based on the Andersen impaction principle. All electronic and moving parts are located outside the critical zone, whilst one stainless steel sampling head for standard 90 to 100 mm Petri dishes collects samples in the containment zone. The Ethernet module provides an interface that is used to communicate with the PC Software and other TCP/IP control software and tools.

Application

MAS-100 ISO NT® Ethernet Module is an additional accessory for Air Sampler that provides an interface that is used to communicate with the PC Software. Microbial air sampling devices are extensively used in Pharmaceutical industries.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service