Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09658

Supelco

n-Dodecane

reference substance for gas chromatography

Synonym(s):

n-Dodecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CH3
CAS Number:
Molecular Weight:
170.33
MDL number:
UNSPSC Code:
12352001
EC Index Number:
203-967-9

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

assay

≥98.5% (GC)

form

liquid

autoignition temp.

200 °C

expl. lim.

0.6 % (v/v)

technique(s)

gas chromatography (GC): suitable

kinematic viscosity

<7 cSt(40 °C)

bp

216.3 °C/1013 hPa

mp

-9.6 °C

transition temp

flash point 85.5 °C

density

0.75 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3

InChI key

SNRUBQQJIBEYMU-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 98.5 % (a/a)
Identity (IR-spectrum): conforms

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

supp_hazards

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_c

70 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service