Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09624

Supelco

Charcoal activated

for gas chromatography 0.5-1 mm (18-35 mesh ASTM)

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
11101522
EC Index Number:
231-153-3

Quality Level

form

solid

technique(s)

gas chromatography (GC): suitable

mp

3550 °C

bulk density

150‑440 kg/m3

storage temp.

15-25°C

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

Charcoal activated for gas chromatography 0.5-1 mm (18-35 mesh ASTM). CAS 7440-44-0, molar mass 12.01 g/mol.

Analysis Note

Resolution: 4.5 - 11.0
Bulk density: 0.4 - 0.5 g/cm³
Zn (Zinc): ≤ 0.002 %
Particle size (0.5 - 1.0 mm): ≥ 70.0 %

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service