Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09085

Millipore

Main charger for MAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403

packaging

pkg of 1 unit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100® Microbial Air Sampler

storage temp.

no temp limit

General description

Mains charger is an additional accessory for microbial air sampler. The MAS-100® is an air sampler, designed to monitor air microbiology in cleanrooms and hygienically ultra-sensitive areas. Its high recovery rate is based on the Andersen impaction principle. An integrated airflow sensor compensates for differences in pressure and temperature in the environment. The Mains charger for MAS-100® has an input voltage of 100 to 240 V AC (47 to 63 Hz, 1.5 to 0.7 A) and output voltage of 24 V DC ± 5% (2.7 A). The cable length varies depending upon the country.

Application

Mains charger is an additional accessory for MAS-100® microbial air sampler that is used for microbial monitoring of ambient air in controlled environments up to ISO 5 cleanrooms. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Linkage

Replaces: 1090850002

Legal Information

MAS-100 is a registered trademark of MBV AG

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service