Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07978

Sigma-Aldrich

Thiourea

special quality for synthesis

Synonym(s):

Thiourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
76.12
MDL number:
UNSPSC Code:
12352300
EC Index Number:
200-543-5
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

Quality Level

form

solid

autoignition temp.

440 °C (Dust)

potency

1750 mg/kg LD50, oral (Rat)
>2800 mg/kg LD50, skin (Rabbit)

pH

6-8 (20 °C, 50 g/L in H2O)

mp

169-173 °C

solubility

137 g/L

density

1.405 g/cm3 at 20 °C

bulk density

640 kg/m3

Storage temp.

no temp limit

SMILES string

S=C(N)N

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

Inchi Key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

General description

special quality for synthesis

Application

Thiourea special quality for synthesis. CAS No. 62-56-6, EC Number 200-543-5.

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service