Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.05650

Supelco

ProteoChrom HPTLC silica gel 60

suitable for HPTLC, plate L × W 20 cm × 10 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

glass support
silica gel 60 matrix

feature

binder Organic Polymer
fluorescent indicator: no

packaging

pkg of 25 plates

technique(s)

HPTLC: suitable

layer thickness

100 μm

plate L × W

20 cm × 10 cm

particle size

5-6 μm

pore size

60 Å medium pore diameter

storage temp.

2-30°C

General description

for peptid analysis 20 x 10 cm

Analysis Note

Specific surface area (according to BET; 5-Pt. measurement): 480 - 540 m²/g
Pore volume (N₂-isotherm): 0.74 - 0.84 ml/g
d 50 (laser diffraction, size distribution): 5 - 7 µm
Layer thickness: 70 - 120 µm
Deviation of layer thickness per plate: ≤ 20 µm
Aminoacid test
hRf-values
- L-Lysine:
2 - 7
- L-Glutaminic acid: 10 - 18
- L Glutamine: 25 - 35
separation number: ≥ 8.5
Typical value determined on an activated plate (120°C, 15 min)
Eluent: 2-Butanol/ pyridine/ ammonia solution 25%/ water (39/34/10/26 v/v/v/v) + 0.05 mol/L Lithium chloride (1mg/ml)

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service