Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.05437

Millipore

MUELLER-HINTON agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

powder

Quality Level

technique(s)

microbiological culture: suitable

pH

7.2-7.6 (37 °C, 35 g/L in H2O, after autoclaving)

solubility

35 g/L

application(s)

clinical
food and beverages
microbiology
pathogen testing

storage temp.

20-25°C

General description

for testing the sensitivity of clinically important pathogens

Analysis Note

Appearance (clearness):clear to opalescent
Appearance (colour):yellowish-brown
pH-value (25 °C):7.2 - 7.6
Solidification behaviour (2 hrs., 40 °C):liquid
Inhibition zone diameter/Ampicillin 10 μg (Escherichia coli ATCC 25922 (WDCM 00013)):16 - 22 mm
Inhibition zone diameter/Tetracyclin 30 μg (Escherichia coli ATCC 25922 (WDCM 00013)):18 - 25 mm
Inhibition zone diameter/Gentamicin 10 μg (Escherichia coli ATCC 25922 (WDCM 00013)):19 - 26 mm
Inhibition zone diameter/Polymyxin B 300 U/IE (Escherichia coli ATCC 25922 (WDCM 00013)):12 - 17 mm
Inhibition zone diameter/SXT 1.25 μg + 23.75 μg (Escherichia coli ATCC 25922 (WDCM 00013)):24 - 32 mm
Inhibition zone diameter/Ampicillin 10 μg (Staphylococcus aureus ATCC 25923 (WDCM 00034)):27 - 35 mm
Inhibition zone diameter/Tetracyclin 30 μg (Staphylococcus aureus ATCC 25923 (WDCM 00034)):19 - 28 mm
Inhibition zone diameter/Gentamicin 10 μg (Staphylococcus aureus ATCC 25923 (WDCM 00034)):19 - 27 mm
Inhibition zone diameter/Polymyxin B 300 U/IE (Staphylococcus aureus ATCC 25923 (WDCM 00034)):7 - 13 mm
Inhibition zone diameter/SXT 1.25 μg + 23.75 μg (Staphylococcus aureus ATCC 25923 (WDCM 00034)):24 - 32 mm
Inhibition zone diameter/Gentamicin 10 μg (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)):16 - 23 mm
Inhibition zone diameter/SXT 1.25 μg + 23.75 μg (Enterococcus faecalis ATCC 33186):≥ 20 mm
Incubation: 24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service