Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.05435

Millipore

MUELLER-HINTON agar acc. to CLSI

for the determination of antibiotic susceptibility including sulfonamides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

powder (dehydrated (DCM))

technique(s)

microbiological culture: suitable

pH

7.1-7.5 (25 °C, 38 g/L in H2O, after autoclaving)

solubility

38 g/L

application(s)

clinical
microbiology

suitability

Enterococcus spp.
Escherichia coli
Pseudomonas spp.
Staphylococcus spp.

General description

for the determination of antibiotic susceptibility including sulfonamides

Analysis Note

Appearance (clearness)opalescent to turbidAppearance (color)brownish-yellowpH-value (25 °C)7.1 - 7.5Growth (Escherichia coli ATCC 25922 (WDCM 00013))good to very goodGrowth (Staphylococcus aureus ATCC 25923 (WDCM 00034))good to very goodGrowth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025))good to very goodGrowth (Enterococcus faecalis ATCC 29212 (WDCM 00087))good to very goodInhibition zone diameter (Escherichia coli ATCC 25922 (WDCM 00013))passes testInhibition zone diameter (Staphylococcus aureus ATCC 25923 (WDCM 00034))passes testInhibition zone diameter (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025))passes testInhibition zone diameter (Enterococcus faecalis ATCC 29212 (WDCM 00087))passes testIncubation: 16-18 hrs.; 35 °C aerobic.Ca (Calcium)Mg (Magnesium)

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service