Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04836

Supelco

Ion exchanger V

(mixed bed exchanger) GR for analysis

Synonym(s):

Mixed bed exchanger

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Quality Level

form

beads

technique(s)

LPLC: suitable

particle size

300-1100 μm

bulk density

650‑700 kg/m3

separation technique

mixed mode

storage temp.

no temp limit

Analysis Note

Appearance: light brown spherules
Exchange capacity (Anionen): ≥ 0.4 mol/l
Exchange capacity (cations): ≥ 0.5 mol/l

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service