Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04306

Supelco

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

vapor pressure

160 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

240 °C

expl. lim.

1.0-8.1 % (v/v)

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C/1013 hPa

mp

-94.3 °C

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.66 g/cm3 at 20 °C

Storage and Stability

Store at +5°C to +30°C.

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-7.6 °F

flash_point_c

-22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service