Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03733

Supelco

StripScan Reference Card

for use with MQuant® nitrate test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

description

semi-quantitative

product line

MQuant®

specific analyte(s)

nitrate

compatibility

for use with MQuant® StripScan App
for use with MQuant® nitrate test

detection method

colorimetric

Storage temp.

15-25°C

General description

for the measurement of MQuant® Nitrate Test, Cat. No.110020 MQuant

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service