Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01347

Millipore

EMB agar

Synonym(s):

Eosin methylene-blue lactose sucrose agar, EMB agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

medium granules

Quality Level

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.0-7.2 (37 °C, 36 g/L in H2O, after autoclaving)

solubility

36 g/L

application(s)

food and beverages
industrial qc
microbiology

storage temp.

20-25°C

suitability

Salmonella spp. (spp.)
Shigella spp. (spp.)

General description

for the detection and isolation of pathogenic Enterobacteriaceae

Analysis Note

Appearance (clearness):clear
Appearance (colour):reddish-brown to violet-brown
pH-value (25 °C):6.9 - 7.3
Growth (Escherichia coli ATCC 25922 (WDCM 00013)):good to very good
Growth (Escherichia coli 194):good to very good
Growth (Escherichia coli ATCC 11775 (WDCM 00090)):good to very good
Growth (Escherichia coli ATCC 23716):good to very good
Growth (Escherichia coli ATCC 8739 (WDCM 00012)):good to very good
Growth (Enterobacter cloacae ATCC 13047 (WDCM 00083)):fair to very good
Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031)):good to very good
Growth (Shigella flexneri ATCC 12022 (WDCM 00126)):good to very good
Growth (Bacillus cereus ATCC 11778 (WDCM 00001)):none to poor
Metallic lustre (Escherichia coli ATCC 25922 (WDCM 00013)):+
Metallic lustre (Escherichia coli 194):+
Metallic lustre (Escherichia coli ATCC 11775 (WDCM 00090)):+
Metallic lustre (Escherichia coli ATCC 23716):+
Metallic lustre (Escherichia coli ATCC 8739 (WDCM 00012)):+
Metallic lustre (Enterobacter cloacae ATCC 13047 (WDCM 00083)):- or poor
Metallic lustre (Salmonella typhimurium ATCC 14028 (WDCM 00031)):-
Metallic lustre (Shigella flexneri ATCC 12022 (WDCM 00126)):-
Metallic lustre (Bacillus cereus ATCC 11778 (WDCM 00001)):-
Incubation: 24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service