Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00641

Millipore

Acrylamide-bis ready-to-use solution 40% (29.1:0.9)

for electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

Quality Level

form

liquid

density

1.03 g/cm3 at 20 °C

storage temp.

2-8°C

Analysis Note

Assay of N,N′-methylene-bisacrylamide (HPLC): 2.8 - 3.2 %
Appearance of solution (colour): almost colourless
Appearance of solution (clearness): almost clear
Free acid (as acrylic acid): ≤ 0.002 %
Spec. Absorptivity A 1%/1cm (290 nm; 1 %; 1 cm; water): ≤ 0.15

pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

target_organs

Peripheral nervous system

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service