Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09-129

Sigma-Aldrich

Anti-SFRP4 Antibody

serum, Upstate®

Synonym(s):

secreted frizzled-related protein 4, FRP-4, sFRP-4, FRPHE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SFRP4(6424)

Specificity

Recognizes SFRP4

Immunogen

Recombinant full length human GST-tagged SFRP4 protein.

Application

Anti-SFRP4 Antibody detects level of SFRP4 & has been published & validated for use in WB.
Immunoblot: 1:500-1:5000 dilution is recommended depending on cell type and method of stimulation.

Optimal working dilutions must be determined by end user.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Routinely evaluated by immunoblot.

Target description

55 kDa

Physical form

Serum

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biao He et al.
Cancer research, 65(3), 743-748 (2005-02-12)
The secreted frizzled-related proteins (SFRPs) function as negative regulators of Wnt signaling and have important implications in tumorigenesis. Frequent promoter hypermethylation of SFRPs has been identified in human cancer. Restoration of SFRP function attenuates Wnt signaling and induces apoptosis in
Secreted frizzled-related protein 4 expression is positively associated with responsiveness to cisplatin of ovarian cancer cell lines in vitro and with lower tumour grade in mucinous ovarian cancers.
Saran, U; Arfuso, F; Zeps, N; Dharmarajan, A
BMC Cell Biology null
Lisa G Horvath et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 10(2), 615-625 (2004-02-05)
Activation of the Wnt-signaling pathway is implicated in aberrant cellular proliferation in a variety of cancers. Secreted frizzled-related protein 4 (sFRP4) is a secreted protein with putative inhibitory activity of the Wnt-signaling cascade through binding and sequestering Wnt ligands. Because
W Roth et al.
Oncogene, 19(37), 4210-4220 (2000-09-12)
Cellular resistance to multiple proapoptotic stimuli and invasion of surrounding brain tissue by migrating tumor cells are main obstacles to an effective therapy for human malignant glioma. Here, we report that the Wnt family of embryonic differentiation genes modulate growth
Taranjit S Gujral et al.
PloS one, 5(4), e10024-e10024 (2010-04-13)
Aberrant Wnt signaling has been implicated in a wide variety of cancers and many components of the Wnt signaling network have now been identified. Much less is known, however, about how these proteins are coordinately regulated. Here, a broad, quantitative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service