Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

07-539-AF488

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys4) Antibody, Alexa Fluor 488 conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

H3K4ac, H3.3B, Lys4 acetylated, H3 histone, family 3A, Lys4 acetylated, H3 histone, family 3B, Lys4 acetylated, Histone H3, Lys4 acetylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

acetylation (Lys4)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3 variants (H3.1, H3.2 and H3.3) have been implicated in the epigenetic memory of cellular state. Genome-wide patterns of H3 are dependent on amino acid sequence and change with cellular differentiation at developmentally regulated loci.

Specificity

Broad species cross-reactivity is expected.
Specificity of this polyclonal antibody toward histone H3 Lys4 acetylation was demonstrated by bead-based multiplex assay and immunogen peptide blocking by Western blotting analysis using the unconjugated antibody (Cat. No. 07-539).

Immunogen

Epitope: H3K4ac
KLH-conjugated synthetic peptide containing the sequence ...RT[Kac]Q... in which [Kac] corresponds to acetylated Lys4 of human histone H3.

Application

Anti-acetyl-Histone H3 (Lys4) Antibody, Alexa Fluor 488 conjugate is an antibody against histone Lys4 acetylation for use in Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected detected nuclear histone H3 Lys4 acetylation in HeLa and A431 cells.
The unconjugated antibody (Cat. No. 07-539) is shown to be suitable also for dot blot, chromatin immunoprecipitation (ChIP), ChIP-seq, multiplex assay, and Western blotting applications.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Immunocytochemistry in HepG2 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected nuclear histone H3 Lys4 acetylation in HepG2 cells.

Target description

~15 kDa observed

Physical form

Protein A purified.
Purified rabbit polyclonal antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service