Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-473-AF647

Sigma-Aldrich

Anti-trimethyl-Histone H3 (Lys4) Antibody, Alexa Fluor 647 Conjugate

from rabbit, ALEXA FLUOR 647

Synonym(s):

H3K4me3, Histone H3 (trimethyl Lys4), H3 histone family, member T, Histone 3, H3, Histone cluster 3, H3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$638.00

$638.00


Estimated to ship onMay 06, 2025


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$638.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$638.00


Estimated to ship onMay 06, 2025


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

vertebrates (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys4)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)

General description

Histone H3 is one of the five main histone proteins that are involved in the structure of chromatin in eukaryotic cells. Histone H3 features a main globular domain and a long N-terminal tail. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3 is specifically phosphorylated during both mitosis and meiosis in patterns that are specifically coordinated in space and time. These phosphorylations are initiated at different phases of the cell division. Histone H3 variants (H3.1, H3.2 and H3.3) have been implicated in the epigenetic memory of cellular state. Genome-wide patterns of H3 are dependent on amino acid sequence and change with cellular differentiation at developmentally regulated loci.

Specificity

Expected to react with most other species based on extreme conserved homology among species.

Immunogen

Epitope: Trimethylated (Lys4)
Synthetic peptide corresponding to residues surrounding and including trimethlyated Lys 4 of Histone H3.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected trimethyl-Histone H3 (Lys4) in NIH/3T3 cells.
The unconjugated version of this antibody has been shown to work in ChIP, DB, WB, ChIP-seq.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-trimethyl-Histone H3 (Lys4) Antibody, Alexa Fluor 647 Conjugate is validated for use in Immunocytochemistry for the detection of trimethyl-Histone H3.

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected trimethyl-Histone H3 (Lys4) in A431 cells.

Target description

~16 kDa observed

Physical form

Affinity purified
Purified rabbit conjugate antibody in buffer containing PBS with 15 mg/ml BSA and 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service