Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-2142

Sigma-Aldrich

Anti-ARHGAP36 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Rho GTPase-activating protein 36, ARHGAP36, RHGXX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

monkey, mouse, rat, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Rho GTPase-activating protein 36 (RHG36 or ARHGAP36) is a GTPase activator for the Rho-type GTPases that converts them to an inactive GDP-bound state. Located in the cytosol, RHG36 is involved in the regulation of small GTPase mediated signal transduction.

Immunogen

GST-tagged recombinant protein corresponding to human ARHGAP36

Application

Anti-ARHGAP36 Antibody is a highly specific rabbit polyclonal antibody, that targets Rho GTPase-activating protein 36 & has been tested in western blotting & ICC.
Immunocytochemisty Analysis: A 1:100 dilution of this antibody detected ARHGAP36 in COS cells (T. Pawson, Mount Sinai Hospital, Toronto.).

Quality

Evaluated by Western Blotting in mouse spleen tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected ARHGAP36 in 10 µg of mouse spleen tissue lysate.

Target description

~68 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service