Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-172-I

Sigma-Aldrich

Anti-FHIT

from rabbit, purified by affinity chromatography

Synonym(s):

Bis(5′-adenosyl)-triphosphatase, AP3A hydrolase, AP3Aase, Diadenosine 5′,5′′′-P1,P3-triphosphate hydrolase, Dinucleosidetriphosphatase, Fragile histidine triad protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... FHIT(2272)

General description

Bis(5′-adenosyl)-triphosphatase (EC 3.6.1.29; UniProt P49789; also known as AP3A hydrolase, AP3Aase, Diadenosine 5′,5′′′-P1,P3-triphosphate hydrolase, Dinucleosidetriphosphatase, Fragile histidine triad protein) is encoded by the FHIT gene (Gene ID 2722) in human. Common fragile sites (CFSs) are large genomic loci that display chromosomal instability in response to genotoxic or replicative stress. Many CFSs harbor genes that contribute to cancer development and are non-randomly altered in cancers through genomic alteration. The Fragile histidine triad (FHIT) gene at FRA3B is one of the earliest and most frequently altered genes in the majority of human cancers. FHIT protein plays a role in apoptosis and prevention of the epithelial-mesenchymal transition (EMT). A FHIT-upregulated microRNA, miR-30c, has been identified to mediate the suppression of EMT and metastasis by directly targeting metastasis genes Metadherin (MTDH), High-mobility group AT-hook 2 (HMGA2), and the mesenchymal markers, Vimentin (VIM) and Fibronectin (FN1), in human lung cancer. In addition, FHIT protein also plays an important role in maintaining genome stability, loss of FHIT protein function leads to nucleotide imbalance, spontaneous replication stress, and DNA breaks. Because FHIT loss-induced DNA damage is checkpoint-independent, cells accumulate further DNA damage during subsequent cell cycles, accruing global genome instability that could facilitate oncogenic mutation acquisition and expedite clonal expansion.

Specificity

This rabbit polyclonal antibody targets a C-terminal epitope region outside the HIT domain.

Immunogen

KLH-conjugated linear peptide corresponding to a C-terminal region sequence of human FHIT.

Application

Anti-FHIT, Cat. No. 07-172-I, is a highly specific rabbit polyclonal antibody that targets FHIT and has been tested in Immunoprecipitation and Western Blotting.
Immunoprecipitation Analysis: 5 µg from a representative lot immunoprecipited FHIT from 500 µg of A549 cell lysate.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected FHIT in 10 µg of A549 cell lysate.

Target description

~17 kDa observed. 16.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 07-172

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service