Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-1381

Sigma-Aldrich

Anti-PP6R3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Serine/threonine-protein phosphatase 6 regulatory subunit 3, SAPS domain family member 3, Sporulation-induced transcript 4-associated protein SAPL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$311.00

$311.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$311.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$311.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP6R3(55291)

General description

PP6R3 (protein Ser/Thr phosphatase 6 regulatory subunit 3), also known as SAPS3 is a regulatory subunit of protein phosphatase 6 which is important for cell survival and is associated with TOR signaling in yeast. PP6R3 has been implicated in the transition from G1 to the S phase of mitosis. The PP6R3 subunit is one of three human versions of SAPs which may also have a role to play in regulation of NF-kappaB.

Immunogen

Histidine-tagged-recombinant protein corresponding to human PP6R3.

Application

Anti-PP6R3 Antibody detects level of PP6R3 & has been published & validated for use in WB, IH(P).
Immunohistochemistry Analysis: 1:200 dilution of a representative lot detected PP6R3 in human kidney and human breast tissues.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1:10,000 dilution of this antibody detected PP6R3 on 10 µg of HeLa cell lysate.

Target description

~130-140 kDa observed. UniProt entry describes 6 isoforms produced by alternative splicing ranging from 88.9 to 98.9 kDa


Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Promising developments in targeted therapies for non-small-cell lung cancer.
Desiree Hao,Manuel Hidalgo
Clinical Lung Cancer null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service