Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

06-598-AF488

Sigma-Aldrich

Anti-acetyl-Histone H4 Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

Histone H4, major, acetyl-Histone H4, AlexaFluor 488 Conjugate, Histone H4, minor, Histone H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Product 06-598-AF488 is not currently sold in your country. Contact Technical Service

biological source

rabbit

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

Tetrahymena sp. (based on 100% sequence homology), protozoa (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

GenBank accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Not specified)

Gene Information

human ... H4C1(8359)
mouse ... H4C1(326619)

General description

Histone H4 (UniProt P02310; P69151; P69152) is encoded by the HHF1 (ORF TTHERM_00498190) and HHF2 (ORF TTHERM_00189170) gene (Gene ID 7823613) in protozoa Tetrahymena. Histone H4 is one of the main histone proteins involved in the structure of chromatin in both prokaryotic and eukaryotic cells. Featuring a main globular domain and a long N terminal tail, H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).

Specificity

Recognizes acetylated histone H4. Cross-reacts with acetylated histone H2B from Tetrahymena and weakly cross-reacts with acetylated histone H2B from HeLa cells. May crossreact with other acetylated proteins.

Immunogen

Epitope: a.a. 2-19
KLH-conjugated peptide corresponding to amino acids 2-19 of Tetrahymena histone H4 (AGG(Ac)KGG(Ac)KGMG(Ac)KVGA(Ac)KRHS-C), acetylated on lysines 5,8,12 and 16.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected acetyl-Histone H4 in HeLa, A431, and NIH/3T3 cells.
The unconjugated antibody (Cat. No. 06-598) is shown to be suitable also for Immunoprecipitation (IP), Chromatin IP (ChIP), and Western blotting applications.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-acetyl-Histone H4 Antibody, AlexaFluor 488 Conjugate is validated for use in Immunocytochemistry for the detection of acetyl-Histone H4.

Quality

Evaluated by Immunocytochemistry in HUVEC cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected acetyl-Histone H4 in HUVECs.

Target description

~11 kDa observed

Physical form

Protein A purified
Purified rabbit antibody conjugate in PBS with 15mg/ml BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service