Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

06-1292

Sigma-Aldrich

Anti-Cep63 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Centrosomal protein of 63 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CEP63(80254)

General description

Centrosomal protein 63 (Cep63) serves an ATM and ATR substrate for spindle assembly activation following chromosomal breakage. It is shown that inactivation of the CEP63 gene in avian DT40 cells impairs spindle assembly and prevents ATM- and ATR-dependent effects on mitosis. Cep63 is also reported to bind to and recruit Cdk1 to centrosomes and is thought to be involved the regulation mitotoic entry, centrosome amplification, and genome maintentance.

Immunogen

Recombinant protein corresponding to human Cep63.

Application

Immunocytochemistry Analysis: A representative lot detected Cep63 in U20S cells. (Nicola Brown, CRUK Clare Hall Laboratories, London, UK)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-Cep63 Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC to detect Cep63 also known as Centrosomal protein of 63 kDa.

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: A 1: 500 dilution of this antibody detected Cep63 in 10 µg of HEK 293 cell lysate.

Target description

~63 kDa observed. Different isoforms or uncharacterized bands may be observed in some endogenous cell lysates. Experiments using transfected cells and siRNA have confirmed detection of Cep63.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK 293 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieyi Xiong et al.
Human molecular genetics, 27(8), 1474-1485 (2018-02-17)
Although splicing is widespread and evolves rapidly among species, the mechanisms driving this evolution, as well as its functional implications, are not yet fully understood. We analyzed the evolution of splicing patterns based on transcriptome data from five tissues of
Denis Fortun et al.
IEEE transactions on medical imaging, 37(5), 1235-1246 (2018-05-05)
The imaging of proteins within macromolecular complexes has been limited by the low axial resolution of optical microscopes. To overcome this problem, we propose a novel computational reconstruction method that yields isotropic resolution in fluorescence imaging. The guiding principle is
Farners Amargant et al.
Molecular human reproduction, 27(11) (2021-09-29)
The mechanism of conversion of the human sperm basal body to a centrosome after fertilization, and its role in supporting human early embryogenesis, has not been directly addressed so far. Using proteomics and immunofluorescence studies, we show here that the
Georgios N Hatzopoulos et al.
Nature communications, 12(1), 3805-3805 (2021-06-23)
Centrioles are evolutionarily conserved multi-protein organelles essential for forming cilia and centrosomes. Centriole biogenesis begins with self-assembly of SAS-6 proteins into 9-fold symmetrical ring polymers, which then stack into a cartwheel that scaffolds organelle formation. The importance of this architecture
Catherine Sullenberger et al.
The Journal of cell biology, 222(12) (2023-09-14)
Centriole duplication is a high-fidelity process driven by Polo-like kinase 4 (Plk4) and a few conserved initiators. Dissecting how Plk4 and its receptors organize within centrosomes is critical to understand the centriole duplication process and biochemical and architectural differences between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service