Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

06-1022

Sigma-Aldrich

Anti-NET34 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

solute carrier family 39 (zinc transporter), member 14, LIV-1 subfamily of ZIP zinc transporter 4, solute carrier family 39 (metal ion transporter), member 14, Zrt-, Irt-like protein 14, Zrt- and Irt-like protein 14, zinc transporter ZIP14, Solute carrie

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

NET34, more commonly known as as Zinc transporter ZIP14, or Zrt/Irt-like protein 14 is a membrane protein. Data suggests this protein functions as a zinc influx transporter and may also be able to transport iron. Three isoforms of this protein have been described.

Specificity

This antibody recognizes NET34 at the extracellular domain.

Immunogen

Epitope: Extracellular domain
KLH-conjugated linear peptide corresponding to human NET 34 at the extracellular domain.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected NET34 in HeLa cells.
Research Category
Epigenetics & Nuclear Function
Use Anti-NET34 Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC to detect NET34 also known as LIV-1 subfamily of ZIP zinc transporter 4, Solute carrier family 39 member 14.

Quality

Evaluated by Western Blot in HEK293T cell lysate.

Western Blot Analysis: 4 µg/mL of this antibody detected NET34 on 10 µg of HEK293T cell lysate.

Target description

~83 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293T cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Wang et al.
Nature medicine, 24(6), 770-781 (2018-06-08)
Patients with metastatic cancer experience a severe loss of skeletal muscle mass and function known as cachexia. Cachexia is associated with poor prognosis and accelerated death in patients with cancer, yet its underlying mechanisms remain poorly understood. Here, we identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service