Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

05-449-AF488

Sigma-Aldrich

Anti-Bin1 Antibody, clone 99D Antibody, Alexa Fluor 488 Conjugate

from mouse, ALEXA FLUOR 488

Synonym(s):

Amphiphysin II, Amphiphysin-like protein, Box-dependent myc-interacting protein 1, Bridging integrator 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$566.00

$566.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$566.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$566.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, mouse, chicken, rat

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

chicken ... Bin1(424228)
human ... BIN1(274)
mouse ... Bin1(30948)

Related Categories

General description

Myc box-dependent-interacting protein 1 (UniProt: O00499; also known as Amphiphysin II; Amphiphysin-like protein; Box-dependent myc-interacting protein 1; Bridging integrator 1, BIN1) is encoded by the BIN1 (also known as AMPHL) gene (Gene ID: 274) in human. BIN1 is expressed in many tissues, but has the highest expression in brain and muscle. In brain it is concentrated in the cytomatrix beneath the plasma membrane of axon initial segments and nodes of Ranvier. In skeletal muscle, it is concentrated around the plasmalemma of T tubules. BIN1 is involved in regulation of synaptic vesicle endocytosis and is also reported to act as a tumor suppressor that inhibits malignant cell transformation. BIN1 is considered as a dominant autoantigen in paraneoplastic Stiff-man syndrome. Mutations in BIN1 gene are known to cause myopathy centronuclear 2 (CNM2), a congenital muscle disorder characterized by progressive muscular weakness and wasting involving mainly limb girdle, trunk, and neck muscles. Ref.: Butler, MH., et al., (1997). J. Cell Biol. 137(6): 1355 1367.

Specificity

Clone 99D, Alexa Fluor 488 Conjugate detects Bin1 in A431 and HeLa cells by immunocytochemistry.

Immunogen

Linear peptide corresponding to human Bin1.

Application

Detect Bin1 using this mouse monoclonal Anti-Bin1 antibody, clone 99D, Alexa Fluor 488 Conjugate, Cat. No. 05-449-AF488, validated for use in Flow Cytometry and Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Bin1 in HeLa cells.

Flow Cytometry Analysis: A 1:400 dilution from a representative lot detected Bin1 in one million HeLa cells.


Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Bin1 in A431 cells.

Target description

64.7 kDa calculated.

Physical form

Protein G purified
Purified mouse monoclonal antibody IgG2b in PBS with 1.5% BSA and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service