Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

05-1951-AF488

Sigma-Aldrich

Anti-trimethyl Histone H3 (Lys27) Antibody, clone 18E9.1, Alexa Fluor 488 Conjugate

clone 18E9.1, from mouse, ALEXA FLUOR 488

Synonym(s):

Histone H3.3, H3F3A, H3.3A, H3F3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

18E9.1, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys27)

Gene Information

human ... H3F3B(3021)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

Linear peptide corresponding to trimethyl Histone H3 (Lys27).

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected trimethyl Histone H3 (Lys27) in NIH/3T3 cells.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
This Anti-trimethyl Histone H3 (Lys27) Antibody, clone 18E9.1, Alexa Fluor 488 Conjugate is validated for use in ICC for the detection of trimethyl Histone H3 (Lys27).

Quality

Evaluated by Immunocytochemistry Analysis in HepG2 cells

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected trimethyl Histone H3 (Lys27) in HepG2 cells.

Alexa Fluor is a registered trademark of Life Technologies.

Target description

17 kDa calculated

Physical form

Protein G Purified
Purified mouse conjugate in buffer containing PBS, 15 mg/ml BSA, and 0.1% Sodium Azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service