Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

05-180-AF647

Sigma-Aldrich

Anti-Cortactin (p80/85) Antibody, clone 4F11, Alexa Fluor 647 Conjugate

clone 4F11, from mouse, ALEXA FLUOR 647

Synonym(s):

Src substrate protein p85, Cortactin, Cortical actin-binding protein, p80, p80/85

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

conjugate

ALEXA FLUOR 647

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F11, monoclonal

species reactivity

hamster, mouse, avian, rat, bovine, human

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CTTN(2017)

General description

Src substrate protein p85 (UniProt Q01406; also known as Cortactin, Cortical actin-binding protein, p80, p80/85) is encoded by the CTTN1 (also known as CTTN, EMS1, P85.25) gene (Gene ID 396455) in Gallus gallus (Chicken) species. Cortactin is a cytosolic protein that promotes polymerization and rearrangement of the actin cytoskeleton, especially the actin cortex around the cellular periphery. Cortactin is activated via phosphorylation by upstream kinases in response to extracellular stimuli. Src is known to phosphorylate cortactin on Tyr421, Tyr466, and Tyr482, leading to cortactin interaction with F-actin and Arp2/3. Erk can also phosphorylate cortactin on Ser405 and Ser418, causing recruitment of additional actin nucleation-promoting factors (NPFs), most importantly N-WASP, to the Arp2/3/ complex. Phosphorylated cortactin thus recruits and stabilizes Arp2/3 complex on preexisting F-actin, providing a nucleation site for a new actin branch to form. Cortactin overexpression in tumor cells enhances invasiveness as a result of highly active lamellipodia formation, also known as invadopodia. Cortactin contains an N-terminal aspartic acid-aspartic acid-tryptophan (DDW) sequence (a.a. 29-31 in chicken; a.a. 20-22 in human) that mediates its interaction with Arp2/3, followed by six and a half repeat sequences (a.a. 80-125, 126-162, 163-199, 200-236, 237-273, 274-310, 311-333 in chicken; a.a. 80-116, 117-153, 154-190, 191-227, 228-264, 265-301, 302-324 in human) that mediates F-actin interaction, a proline-rich coiled coil domain (a.a. 349-410 in chicken; a.a. 348-401 in human) that mediates Src SH3 domain interaction, and a C-terminal SH3 domain (a.a. 505-563 in chicken; a.a. 492-550 in human) that contain Erk phosphorylation sites and is important for cortactin interaction with NPFs other than Arp2/3.

Specificity

Recognizes Cortactin.

Immunogen

Mixture of affinity-purified tyrosine phosphoproteins from chick embryo fibroblasts expressing activated pp60src.

application

Anti-Cortactin (p80/85) Antibody, clone 4F11, Alexa Fluor 647 Conjugate is an antibody against Cortactin for use in Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Cortactin (p80/85) in NIH/3T3 cells.
The unconjugated antibody (Cat. No. 05-180) is shown to be suitable also for immunocytochemistry, immunofluorescence, immunoprecipitation, and Western blotting applications.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Cortactin (p80/85) in A431 cells.

Target description

~62 kDa observed.

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaushik Pal et al.
The Journal of cell biology, 220(7) (2021-04-28)
Podosomes and invadopodia, collectively termed invadosomes, are adhesive and degradative membrane structures formed in many types of cells and are well known for recruiting various proteases. However, another major class of degradative enzymes, deoxyribonuclease (DNase), remains unconfirmed and not studied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service