Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W-003

Supelco

Warfarin solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

CC(=O)CC(C1C(=O)Oc2ccccc2C1=O)c3ccccc3

InChI

1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,17H,11H2,1H3

InChI key

FQEPJUOLUDFINX-UHFFFAOYSA-N

General description

Warfarin is an anticoagulant sold under the trade names Coumadin® and Jantoven®. Warfarin is also used a pesticide, most often as rat poison. This Snap-N-Spike® reference solution is applicable for use in clinical toxicology, forensic analysis, therapeutic drug monitoring or environmental monitoring and diagnostic testing applications by LC-MS/MS or GC/MS

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Coumadin is a registered trademark of Bristol-Myers Squibb Co.
Jantoven is a registered trademark of Upsher-Smith Laboratories, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1A - STOT RE 2

target_organs

Blood

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ask the doctors. I thought one of the reasons to receive a heart valve made from cow tissue was that you don't have to take blood thinners. Now my doctor is telling me that I will need to take warfarin [Coumadin] for several months after I have my aortic valve replaced. Has the thinking changed?
Thomas Lee et al.
Harvard heart letter : from Harvard Medical School, 23(7), 2-2 (2013-08-07)
Rapid resolution of acute subdural hematoma in a coagulopathic patient.
Navjot Chaudhary et al.
The Canadian journal of neurological sciences. Le journal canadien des sciences neurologiques, 40(4), 599-600 (2013-06-22)
Continuing antithrombotics advised for stroke survivors during certain procedures.
Mike Mitka
JAMA, 309(24), 2537-2537 (2013-06-27)
[Vascular calcification: a common but overlooked condition].
Peter Stenvinkel
Lakartidningen, 110(21), 1041-1044 (2013-06-29)
Choosing a treatment for deep venous thrombosis and pulmonary embolism.
Prescrire international, 22(138), 131-131 (2013-07-03)

Articles

Comparative analysis of Supel™ BioSPME 96-Pin device with a rapid equilibrium dialysis technique for accuracy of measured values, sample cleanliness, and workflow time in drug protein binding.

Protocols

This study demonstrates the analysis of Warfarin in plasma samples utilizing chiral and achiral (reversed-phase) LC-MS and effective sample prep to remove endogenous phospholipids

HPLC Analysis of Warfarin™ Anticoagulant on Ascentis® C18

Chromatograms

suitable for GCsuitable for GCsuitable for GCsuitable for GCShow More

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service