Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V-059

Supelco

beta-Carotene(10,10′,11, 11′)-13C4 solution

100 μg/mL (70:30 THF:Ethanol with 0.1% BHT (w/v)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3613C4H56
Molecular Weight:
540.84
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (70:30 THF:Ethanol with 0.1% BHT (w/v))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

Storage temp.

−70°C

General description

This Snap-N-Shoot® stable-labeled internal standard is suitable for quantification of β-carotene levels in serum or other matrices by LC/MS or GC/MS for clinical diagnostic, or food and nutrition testing applications. β-Carotene, also known as E160a when used as food coloring supplement, is a precursor to vitamin A. Serum levels of β-carotene are an adequate measurement of clinical status and can confirm the diagnosis of carotenoderma and is used to detect fat malabsorption. Low levels may be found in cases of steatorrhea.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-6.2 °F - closed cup

flash_point_c

-21.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service