Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCK-001

Supelco

Chlordecone

99%, vial of 1 g, (Kepone equiv.), Cerilliant®

Synonym(s):

Kepone, Chlordecone, Perchloropentacyclo[5.3.0.02,6.03,9.04,8]decan-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10Cl10O
CAS Number:
Molecular Weight:
490.64
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

assay

99%

packaging

vial of 1 g

manufacturer/tradename

Cerilliant®

application(s)

environmental
forensics and toxicology

format

neat

storage temp.

room temp

SMILES string

ClC1(Cl)C2(Cl)C3(Cl)C4(Cl)C(=O)C5(Cl)C3(Cl)C1(Cl)C5(Cl)C24Cl

InChI

1S/C10Cl10O/c11-2-1(21)3(12)6(15)4(2,13)8(17)5(2,14)7(3,16)9(6,18)10(8,19)20

InChI key

LHHGDZSESBACKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Huff et al.
Journal of environmental pathology and toxicology, 1(4), 377-395 (1978-03-01)
This overview provides a narrative survey of the reference literature on Kepone. The circumstances surrounding the "Hopewell incident" have increased the awareness and vigilance of state and federal agencies regarding human and environmental exposure to toxic chemicals. Highlights of this
Comparative toxicology of chlordecone (Kepone) in humans and experimental animals.
P S Guzelian
Annual review of pharmacology and toxicology, 22, 89-113 (1982-01-01)
H M Mehendale
Journal of toxicology and environmental health, 8(5-6), 743-755 (1981-11-01)
Chlordecone (Kepone) is a decachloroketone analog of the dodecachlorohydrocarbon mirex and is used as a stomach poison insecticide. Despite the structural similarity to mirex, chlordecone is unlike mirex in general organ-specific toxic properties. Chlordecone is primarily accumulated in the liver
M D Reuber
Journal of environmental pathology and toxicology, 2(3), 671-686 (1979-01-01)
Kepone is unmistakably carcinogenic in rats and mice. Kepone induced malignant tumors in the liver of rats and mice in the NCI studies and in the liver in rats in the Medical College of Virginia study. Malignant tumors were also
S B Rao et al.
Indian journal of biochemistry & biophysics, 30(4), 191-198 (1993-08-01)
Why is a low dose of toxic chemical nontoxic? What makes a larger dose of the same chemical toxic? Extensive work done to understand the mechanism of halomethane hepatotoxicity and its potentiation by chlorinated insecticide, chlordecone has resulted in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service