SCD-152
Dibenzo[a,h]pyrene
vial of 10 mg, analytical standard, Cerilliant®
About This Item
grade
analytical standard
packaging
vial of 10 mg
manufacturer/tradename
Cerilliant®
application(s)
environmental
forensics and toxicology
format
neat
storage temp.
room temp
SMILES string
c1ccc2c(c1)cc3ccc4c5ccccc5cc6ccc2c3c46
InChI
1S/C24H14/c1-3-7-19-15(5-1)13-17-9-12-22-20-8-4-2-6-16(20)14-18-10-11-21(19)23(17)24(18)22/h1-14H
InChI key
RXUSYFJGDZFVND-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- In environmental samples using liquid chromatography–atmospheric pressure laser ionization–mass spectrometry (LC-APLI-MS).[3]
- In spiked smoke flavorings[2] and baby food products[1] using gas chromatography coupled to mass spectrometry (GC-MS) technique.
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Carc. 1B - Muta. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service