Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCB-002

Supelco

Benzo[b]fluoranthene

vial of 1 g, analytical standard, Cerilliant®

Synonym(s):

2,3-Benzfluoranthene, 3,4-Benz[e]acephenanthrylene, 3,4-Benzfluoranthene, 3,4-Benzofluoranthene, Benz[e]acephenanthrylene, Benzo[e]fluoranthene, NSC 89265

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

vial of 1 g

manufacturer/tradename

Cerilliant®

mp

163-165 °C (lit.)

application(s)

environmental
forensics and toxicology

format

neat

storage temp.

room temp

SMILES string

c1ccc-2c(c1)-c3cccc4c3c-2cc5ccccc45

InChI

1S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12H

InChI key

FTOVXSOBNPWTSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeh-Chung Chien et al.
Archives of toxicology, 84(10), 767-776 (2010-06-24)
Although urinary 1-hydroxypyrene (1-OHP) is the most relevant parameter for assessing exposure to polycyclic aromatic hydrocarbons, the inability to further elucidate the intra- and inter-individual variability, specificity and kinetics makes it difficult to enhance its value as an exposure predictor.
Vapor phase and particulate bound polycyclic aromatic hydrocarbons in the smoke of mosquito coils.
J M Lin et al.
Bulletin of environmental contamination and toxicology, 59(6), 868-874 (1997-12-24)
Hitoshi Ishikawa et al.
Cancer letters, 240(1), 29-35 (2005-10-26)
Air pollution has been suggested to cause genetic damage from investigations of many biological markers that measure cytogenetic damage in humans. Here, we evaluated the genotoxic effects of ambient air pollution by investigating the extent of cytogenetic damage in human
M J Mass et al.
Carcinogenesis, 17(8), 1701-1704 (1996-08-01)
The polycyclic aromatic hydrocarbon benzo[b]fluoranthene (B[b]F) is a pervasive constituent of environmental combustion products. We sought to examine the lung tumorigenic activity of B[b]F in strain A/J mice, to study the relationship between formation and decay of B[b]F-DNA adducts and
S Nesnow et al.
Environmental health perspectives, 101 Suppl 3, 37-42 (1993-10-01)
DNA-carcinogen adducts offer a potential dosimeter for environmental genotoxicants reaching the exposed individual. Because the target tissues for many chemical carcinogens are not readily accessible for monitoring adducts in humans, peripheral blood lymphocytes (PBLs) have served as surrogate sources of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service