Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S-070

Supelco

3′-Hydroxystanozolol-D3

vial of 5 mg, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H29D3N2O2
CAS Number:
Molecular Weight:
347.51
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

vial of 5 mg

manufacturer/tradename

Cerilliant®

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

format

neat

Storage temp.

−20°C

SMILES string

OC1=NNC2=C1C[C@@]3(C)[C@](CCC4C3CC[C@@]5(C)C4CC[C@@]5(O)C([2H])([2H])[2H])([H])C2

InChI

1S/C21H32N2O2/c1-19-11-14-17(22-23-18(14)24)10-12(19)4-5-13-15(19)6-8-20(2)16(13)7-9-21(20,3)25/h12-13,15-16,25H,4-11H2,1-3H3,(H2,22,23,24)/t12-,13?,15?,16?,19-,20-,21-/m0/s1/i3D3

Inchi Key

SWPAIUOYLTYQKK-XALPJPGDSA-N

General description

3′-Hydroxystanozolol is a major urinary metabolite of stanozolol, an anabolic androgenic steroid. Stanozolol is used as a treatment for osteoporosis and steroid-deficiency but has also been abused in sports as a performance-enhancing drug. This internal standard is suitable for quantitation of 3′-Hydroxystanozolol levels in urine, serum, or plasma by LC/MS or GC/MS for sports testing, urine drug testing, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service